Anti-6xHis Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-382 600-401-382
CA600-401-382EA 574.99 CAD
RL600-401-382 CA600-401-382
Anti-6xHis Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-6X HIS EPITOPE TAG Reacts with Pan

Type: Tag
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR