JAW™ Ready-to-Use Dry Buffer Packs, G-Biosciences
Supplier: G-Biosciences
G-Biosciences' Dry Powder Buffer Packs are easy-to-use, pre-mixed buffer powders that simply require adding water to generate for use.
G-Biosciences' JAW™ Phosphate Buffered Saline (PBS) packs [1X] each make one liter of: 2.7mM potassium chloride, 127mM sodium chloride and 10mM phosphate buffer (at a pH between 7.3 and 7.5). Each JAW™ PBS [10X] pack makes one liter of: 27mM potassium chloride, 1.37M sodium chloride and 0.1M phosphate buffer (at a pH between 7.3 and 7.5).
G-Biosciences' JAW™ Tris Buffered Saline [1X] packs each make one liter of: 25mM Tris, 140mM sodium chloride, and 3mM potassium chloride (at a pH between 7.25 and 7.55). Each JAW™ TBS [20X] pack makes one liter of: 0.5M Tris, 2.8M sodium chloride, and 60mM potassium chloride (at a pH between 7.25 and 7.55).
G-Biosciences also offers readymade solutions of PBS and TBS (10X concentrated) in convenient cubes or various sizes.
Ordering information: Accessories available