Nalgene® Diagnostic Bottles, Translucent Amber, HDPE, with Closure, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
312084-9125 312084-9025 312084-9050
16059-654CS 1894.74 CAD
16059-654 16059-662 16059-670
Nalgene® Diagnostic Bottles, Translucent Amber, HDPE, with Closure, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene translucent amber high density polyethylene diagnostic bottles come with opaque amber polypropylene closures.
These bottles are used in test kits and many small-volume applications. Diagnostic bottles can be used for sample storage and shipping of small volume, valuable reagents that are light sensitive.
Ordering information: Bulk packed for packaging applications.