Nalgene® Sealing Cap Assemblies, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS3131-0020 DS3131-0024
21009-864CS 99.23 CAD
21009-864 21009-886
Nalgene® Sealing Cap Assemblies, Thermo Scientific
Closures Bottle Closures
Nalgene Sealing cap Assemblies seal Nalgene Oak Ridge centrifuge tubes and bottles
Cat. No. DS3131 features shoulder support ring to prevent shifting, binding or collapse of bottle neck. Before autoclaving, set closure on top of the container without engaging the threads.
Recommended for use at maximum speeds (10,000 x g or greater) without leakage, even when full. Also, recommended for spinning hazardous materials compatible with centrifuge tube material.
Order Now
SPECIFICATIONS
Environmentally Preferable