Human Normal Cell Line Blots, G-Biosciences
Supplier: G-Biosciences
TB50 TB51 TB52 TB53
CA82023-230EA 4008.74 CAD
CA82023-230 CA82023-232 CA82023-234 CA82023-236
Human Normal Cell Line Blots, G-Biosciences
Blotting Kits
G-Biosciences offers ready-to-screen, normal tissue human cell line blots from epithelial, endothelial, skin, and muscle tissue.
G-Biosciences' premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
G-Biosciences' premade blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact ourMore Product Information