Brain Tissue Region Single Species Blot, G-Biosciences
Supplier: G-Biosciences
TB40 TB41 TB57 TB59
CA82023-210EA 5282.34 CAD
CA82023-210 CA82023-212 CA82023-246 CA82023-248
Brain Tissue Region Single Species Blot, G-Biosciences
Blotting Kits
G-Biosciences offers ready-to-screen, normal tissue, brain tissue region blots
G-Biosciences' premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
G-Biosciences' premade blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact our technical department for the latest information on the blots as G-Biosciences reserves the right to change the blot profile due to the availability of GenLysate™.
The brain tissue specific regions are prepared by carefully dissecting out anatomically and functionally distinct regions of adult mouse, rat, human, and primate brains.
Ordering information: Accessories available.