Nalgene Suspended Magnetic Stir Bars, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS6630-0250 DS6630-1000 DS6630-4000
58948-782CS 256.13 CAD
58948-782 58948-821 58948-864
Nalgene Suspended Magnetic Stir Bars, Thermo Scientific
Stirrers Magnetic Stirring Bars
Nalgene Floating Magnetic Stir Bars are designed for constant low-speed stirring action in tissue culture applications
The stir bars are ideal for tissue culture applications. The bar is slightly raised off the bottom surface of the vessel to minimize grinding effect on cells