Human Single Tissue Single Species Blot, Normal Tissue, G-Biosciences
Supplier: G-Biosciences
TB01 TB04 TB07 TB10 TB13 TB16 TB19 TB22 TB25 TB60 TB61 TB62 TB63 TB64 TB65 TB66 TB67 TB68 TB69 TB70 TB71
CA82023-130EA 1479.2 CAD
CA82023-130 CA82023-136 CA82023-142 CA82023-148 CA82023-154 CA82023-160 CA82023-166 CA82023-172 CA82023-178 CA95029-338 CA95029-340 CA95029-342 CA95029-344 CA95029-346 CA95029-348 CA95029-350 CA95029-352 CA95029-354 CA95029-356 CA95029-358 CA95029-360
Human Single Tissue Single Species Blot, Normal Tissue, G-Biosciences
Blotting Kits
G-Biosciences offers a selection of ready-to-screen, human, single tissue blots from normal tissues.
  • Blots are ready to be blocked and probed with the antibodies of choice
  • Each blot contains two protein lanes, a protein marker lane and a lane with GenLysate™ sample
G-Biosciences premade western blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
Ordering information: Accessories available.