White Polypropylene Screw Caps, Unlined, Qorpak®
Supplier: Qorpak
CAP-00387 CAP-00366 CAP-00356 CAP-00393 CAP-00398 CAP-00380 CAP-00403 CAP-06638 CAP-05088 CAP-06640 501749
16196-306CS 576.38 CAD
16196-306 16196-308 16196-311 16196-318 16196-320 16196-324 16196-326 89080-564 89080-566 89239-416 76236-112
White Polypropylene Screw Caps, Unlined, Qorpak®
Closures Bottle Closures
White Unlined Polypropylene Caps are known for good impact strength, cost effectiveness, and pliability.
  • Autoclavable
  • Suitable for dry products
When autoclaving, place cap loosely on top of jar.