White Metal Screw Caps, Pulp/Polyethylene Liner, Qorpak®
Supplier: Qorpak
CAP-06659 CAP-05015 CAP-06664 CAP-06662 CAP-05016 CAP-06660 CAP-00440 CAP-00435 CAP-00453 CAP-00464 CAP-00458 CAP-00419
11310-804CS 1707.08 CAD
11310-804 11311-150 11311-152 11311-154 16198-794 16198-796 16218-110 16087-200 89204-934 89204-936 89204-940 16198-788 76535-006 76535-008 76535-010 89204-937 89204-941
White Metal Screw Caps, Pulp/Polyethylene Liner, Qorpak®
Closures Bottle Closures
Pulp/Polyethylene lined metal caps are a great general-purpose closure.
They are ideal for powders and non-acidic food products and consumer products such as jar candles and honey.
Order Now
SPECIFICATIONS
Environmentally Preferable