Qorpak® Black Phenolic Screw Caps, Pulp/Aluminum Foil Liner
Supplier: Qorpak
CAP-00110 CAP-00124 CAP-00162 CAP-00213 CAP-00266 CAP-00278 CAP-00290 CAP-00301 CAP-00312 CAP-00285 CAP-00307
11311-288PK 33.58 CAD
11311-288 11311-292 11311-296 11311-298 11311-302 11311-304 11311-306 16087-212 16087-214 16087-218 16087-220 16087-222 16087-224 16087-226 16087-228 16087-230 16087-232 16087-234 16087-236 16087-238 89204-930 76169-366
Qorpak® Black Phenolic Screw Caps, Pulp/Aluminum Foil Liner
Closures Bottle Closures
Pulp/Aluminum Foil lined caps are typically used for food applications as they will not cause odor or taste problems
They can be used for non-acid, non-alkaline products. Great for organic solvents, chrome cleaners, brake fluids and mineral oils.