Black Phenolic Screw Caps, F217 and Fluoropolymer Resin Liner, Qorpak®
Supplier: Qorpak
CAP-06690 CAP-00145 283051 CAP-00171 CAP-00191 CAP-00205 CAP-00222 CAP-00276 CAP-00310
71001-586CS 1155.7 CAD
71001-586 16151-899 76209-746 76500-058 76539-310 76539-312 76539-314 76539-316 76539-320 76539-328
Black Phenolic Screw Caps, F217 and Fluoropolymer Resin Liner, Qorpak®
Closures Bottle Closures
F217 and PTFE liners resist attack from virtually all chemicals at room temperature
The F217 features a low density foam core between two LDPE discs and are applied to the back of a PTFE liner. This provides a tight seal with a low moisture transmission rate. Analytical lab samples, high purity chemicals, strong acids, solvents, environmental samples, pharmaceuticals, and diagnostic reagents.