Nunc® EasyFill Cell Factories, Nunclon Delta Surface, Thermo Scientific
Supplier: Thermo Fisher Scientific
140410 140000 140250 140360 140400 140000 140250 140360 140400 140410
89106-780CS 3267.87 CAD
89106-780 89089-300 89089-302 89089-304 89089-306 CA89089-300 CA89089-302 CA89089-304 CA89089-306 CA89106-780
Nunc® EasyFill Cell Factories, Nunclon Delta Surface, Thermo Scientific
Bioreactors
Highly versatile and easy to use, the EasyFill Cell Factory system offers a sterile, single-use solution designed to optimize valuable incubator space and reduce repetitious manual handling
The EasyFill Cell Factory is easy to use. Simply pour media directly into cell factory, equilibrate by placing cell factory on side with small port, turn 90° so the filling inlet is up, and the medium will be separated with equal amounts in each chamber. With filling the inlet up, carefully tilt the cell factory to a horizontal incubation position. Incubate following the appropriate protocol. Loosen cap and empty directly into reservoir.
Suitable for both research and commercial-scale production.
Ordering information: Accessories (sold separately) include Start-up Kit (Part No. 170769), Filter with Connector (Part No. 173248), Air Filter with Connector (Part No. 173249), and Filter Cap (Part No. 146008).