Packer Bottles, PET, Clear, Wide Mouth, Qorpak
Supplier: Qorpak
PLC-06877 PLC-06996 PLC-06836 PLC-06762 PLC-07300 274632 PLC-08925 PLC-11297 PLC-06837 PLC-06838 PLC-07520 PLC-09008
11311-640CS 650.18 CAD
11311-640 89234-738 30617-180 11311-636 11311-639 75880-446 76487-250 76487-252 76487-254 76487-256 76487-360 76487-362 76487-364 76487-366
Packer Bottles, PET, Clear, Wide Mouth, Qorpak
Bottles Roller Bottles
PET Packer Bottles feature glass like clarity and are durable and rigid.
  • PET packers are stress and impact resistant
  • PET bottles are ideal for oil analysis
They have exceptional gas and moisture barrier properties and a good alcohol and solvent barrier.
Ordering information: Bottle 30617-180 is Vacuum and Ionized. Bottle 89234-738 is Super Cleaned for Oil Analysis.
Order Now
SPECIFICATIONS
Environmentally Preferable