Lab Style Bottles, High-Density Polyethylene, Narrow Mouth, Qorpak®
Supplier: Qorpak
PLA-03149 PLA-03153 PLA-03155 PLA-03157 PLA-03159 501138 249619
16086-016CS 164.17 CAD
16086-016 16086-028 16086-032 16086-048 16086-060 93001-664 80080-922 76003-538 76289-364
Lab Style Bottles, High-Density Polyethylene, Narrow Mouth, Qorpak®
Bottles Plastic Bottles
Narrow mouth HDPE lab style bottles are supplied with linerless polypropylene caps.
Excellent chemical resistance and the leak resistant closure make these ideal for storing, shipping, and packaging liquids for long periods. HDPE bottles are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping. Linerless white polypropylene caps offer excellent impact strength, cost effectiveness, and pliability. Caps attached to the convenience pack and enclosed on the bulk pack