Standard Wide Mouth Bottles, Vacuum and Ionized, Clear
Supplier: Qorpak
GLC-01773 GLC-01836 GLC-01861 GLC-01885 GLC-01746 GLC-01759 GLC-01790
11311-402CS 156.76 CAD
11311-402 11311-478 36319-744 36319-790 36319-791 36319-792 36319-793 36319-794 89204-696 89204-698 76487-370 76487-372 76487-374
Standard Wide Mouth Bottles, Vacuum and Ionized, Clear
Bottles Glass Bottles
The vacuum and ionized cleaning process removes contaminants such as loose dirt, carton lint, aerosols, and fine glass particles.
  • Extremely cost-effective
  • No need to waste valuable time rewashing bottles
  • Provides a clean final appearance
  • Seals cleanliness into each bottle
  • Each container is individually cleaned, and will not collect dust, flies, and other debris while stored in the lab
Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Then the air passes through an absorbent filter to remove gases. A vacuum then draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. The air inside the bottle will not interfere with the product in any way.