French Square Bottles, Vacuum and Ionized, Clear, Wide Mouth
Supplier: Qorpak
GLC-01393 GLC-01369 GLC-01353 GLC-01243 GLC-01287 GLC-01314 GLC-01339 GLC-01367 GLC-01319 GLC-01372 GLC-01322 GLC-01346 GLC-06158 GLC-01267 GLC-01292
11311-418CS 405.81 CAD
11311-418 16151-986 16218-278 36319-711 36319-712 36319-713 36319-714 36319-715 36319-755 36319-756 36319-757 36319-758 36319-759 36319-760 36319-761 36319-800 36319-801 36319-802 89201-532 89204-872 89204-874 76041-652 76487-308 76487-310 76487-314 76487-316 76487-318 76487-320 76487-322 76487-324 76487-326
French Square Bottles, Vacuum and Ionized, Clear, Wide Mouth
Bottles Glass Bottles
The vacuum and ionized cleaning process removes contaminants such as loose dirt, carton lint, aerosols, and fine glass particles.
  • Extremely cost-effective
  • No need to waste valuable time rewashing bottles
  • Provides a clean final appearance
  • Seals cleanliness into each bottle
  • Each container is individually cleaned, and will not collect dust, flies, and other debris while stored in the lab
Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Then the air passes through an absorbent filter to remove gases. A vacuum then draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. The air inside the bottle will not interfere with your product in any way.