Boston Round Bottles, Vacuum and Ionized, Clear, Narrow Mouth
Supplier: Qorpak
GLC-01227 GLC-01096 GLC-01140 GLC-01175 GLC-01207 GLC-01085 GLC-01102 GLC-01126 GLC-01188 GLC-01220 GLC-01133 GLC-01167 GLC-01232 GLC-01165 GLC-01154 GLC-01195 GLC-01122 GLC-01148 GLC-01182
11311-466CS 77.16 CAD
11311-466 11311-468 36319-717 36319-719 36319-720 36319-721 36319-763 36319-764 36319-765 36319-767 36319-768 89031-216 89031-218 89031-220 89031-222 89204-854 11311-465 10818-468 76487-282 76487-284 76487-286 76487-288 76487-290 76487-292 76487-294 76487-296 76487-298 76487-300
Boston Round Bottles, Vacuum and Ionized, Clear, Narrow Mouth
Bottles Glass Bottles
The vacuum and ionized cleaning process removes contaminants such as loose dirt, carton lint, aerosols, and fine glass particles.
  • Extremely cost-effective
  • No need to waste valuable time rewashing bottles
  • Provides a clean final appearance
  • Seals cleanliness into each bottle
  • Each container is individually cleaned, and will not collect dust, flies, or other debris while stored in the lab
Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Then the air passes through an absorbent filter to remove gases. A vacuum then draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. The air inside the bottle will not interfere with your product in any way.