RCX-30 Anion Exchange HPLC Columns, Hamilton Company

Supplier: Hamilton
79370 79371 79372
89221-082EA 1296.11 CAD
89221-082 89221-084 89221-086
RCX-30 Anion Exchange HPLC Columns, Hamilton Company
Chromatography Columns
Hamilton Company offers one of the most comprehensive selections of chromatography columns in the industry.

  • pH Stable from 1 to 13
  • Isocratic Separation of Mono and Disaccharides
  • Isocratic or Gradient Modes
  • Higher Ion Exchange Capacity Support for Separation of Closely Related Carbohydrates
  • Compatible with PAD and RI Detectors
  • Custom Hardware Dimensions Available
  • Buffer strength 0.0 to 1.0N

Each column is manufactured under rigorous criteria and tested for resolution and retention time accuracy.

RCX-30 carbohydrate analysis columns provide longerMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR