PRP-X600 DNA Analysis HPLC Columns, Hamilton Company

Supplier: Hamilton
79361 79360 79362
21493-096EA 499.21 CAD
21493-096 21493-097 89221-074
PRP-X600 DNA Analysis HPLC Columns, Hamilton Company
Chromatography Columns
Hamilton Company offers one of the most comprehensive selections of chromatography columns in the industry.

  • Limited Porosity Allows Separation of Large DNA Fragments
  • pH Stable from 1 to 13
  • Higher Sample Capacity than Nonporous Supports
  • Guard Column in PEEK
  • Buffer strength 0.0 - 1.0N

Each column is manufactured under rigorous criteria and tested for resolution and retention time accuracy.

The unique porosity provides fast separation with better sample capacity than non-porous supports. Polymeric weak-base anion exchange packing for gradientMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Max. pressure 5000 psi 5000 psi 5000 psi
Temperature limit pH 1 to 7.9 (5 to 60°C); pH 8 to 13 (5 to 30°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR