Square Hedpaks®, High-Density Polyethylene, Qorpak®
Supplier: Qorpak
PLC-14633 PLA-03326
89030-124CS 4357.29 CAD
89030-124 89166-156 89166-158 CAPLA-05975
Square Hedpaks®, High-Density Polyethylene, Qorpak®
Bottles Carboys
The Hedpak® is a rigid, high-density polyethylene container.
  • The square design saves space when storing and shipping
An integral handle on the top face with molded pouring vent, along with a finger recess on the bottom of the bottle make it easy-to-use.
Order Now
SPECIFICATIONS
Environmentally Preferable