XIT™ DNA for Protein-Free DNA Isolation, G-Biosciences
Supplier: G-Biosciences
786-301 786-290 786-294 786-295 786-296 786-297 786-298 786-303 786-304 786-337 786-338 786-339 786-340 786-341 786-342 786-343 786-344 786-345 786-346 786-347 786-348 786-349 786-350
CA89167-750EA 2072.94 CAD
CA89167-750 CA95057-626 CA95057-628 CA95057-630 CA95057-632 CA95057-634 CA95057-636 CA95057-638 CA95057-640 CA95057-642 CA95057-644 CA95057-646 CA95057-648 CA95057-650 CA95057-652 CA95057-654 CA95057-656 CA95057-658 CA95057-660 CA95057-662 CA95057-664 CA95057-666 CA95057-668
XIT™ DNA for Protein-Free DNA Isolation, G-Biosciences
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
G-Biosciences offers a wide selection of genomic isolation kits that purify high quality genomic DNA from a variety of sources and for a wide array of applications. G-Biosciences' XIT™ DNA kits produce protein-free, high quality DNA through the principle of cell lysis, protein digestion and precipitation, and genomic DNA purification. No chloroform or phenol extraction is required. High quality DNA can be isolated from sample types including: animal tissues, cells, whole blood, bacteria, Buccal cells, plant tissues, mouse tail, yeast, and FFPE tissue. G-Biosciences' XIT™ DNA kit procedures remove contaminants and enzyme inhibitors, allowing the purified DNA to be ready for immediate use for all downstream analyses. The purified DNA from G-Biosciences' XIT™ DNA kits have a A₂₆₀/A₂₈₀ ratio, between 1.7 and 1.9 (with the exception of the Buccal cells kit with a ratio between 1.8 and 2.0), and are up to 200 kb in size.
G-Biosciences' XIT™ Genomic DNA kits for Animal/Plant/FFPE Tissues, Mouse Tail, and Buccal Cells all use a three main step procedure that includes: rapid and efficient cell lysis then enzymatic protein digestion and complete protein precipitation, followed by DNA precipitation.
G-Biosciences' XIT™ Genomic DNA from Tissue kit is designed for the isolation of genomic DNA from fresh, frozen or methanol/acetone fixed tissues. The kit is supplied with three protocols for the isolation of: 1 - 10 mg fresh or frozen tissue, 50 - 100 mg fresh or frozen tissue or 5 - 10 mg fixed tissue.
G-Biosciences' XIT™ Genomic DNA from Plant Tissue kit is designed for the isolation of genomic DNA from fresh or frozen plant tissues.
G-Biosciences' XIT™ Genomic DNA from FFPE Tissue kit is designed for the isolation of genomic DNA from formalin fixed, paraffin embedded tissue.
G-Biosciences' XIT™ Genomic DNA from Mouse Tail kit is designed for the isolation of genomic DNA from 5 mm sections of mouse tail.
G-Biosciences' XIT™ Genomic DNA from Blood kit is designed for the isolation of genomic DNA from blood, bone marrow and buffy coat. The kit uses four main steps that are red blood cell lysis, rapid & efficient nuclear lysis, complete protein precipitation followed by DNA precipitation.
G-Biosciences' XIT™ Genomic DNA from Cells kit is designed for the isolation of genomic DNA from adherent or suspension cultured cells. The kit uses rapid & efficient nuclear lysis and complete protein precipitation, followed by DNA precipitation.
G-Biosciences' XIT™ Genomic DNA from Gram Negative Bacteria kit is designed for the isolation of genomic DNA from overnight cultures. XIT™ Genomic DNA from Gram Negative Bacteria kit protocol is designed to use 1 ml overnight culture; however the protocol can be easily adapted for larger tissue sample sizes. The kit uses three main steps that are rapid & efficient bacterial lysis, complete protein precipitation, followed by DNA precipitation.
G-Biosciences' XIT™ Genomic DNA from Gram Positive Bacteria kit is designed for the isolation of genomic DNA from overnight cultures. The kit uses three main steps that are rapid & efficient enzymatic digestion of cell walls, spheroplast lysis and complete protein precipitation, followed by DNA precipitation.
G-Biosciences' XIT™ Genomic DNA from Yeast kit is designed for the isolation of genomic DNA from overnight yeast cultures. The kit uses three main steps that are rapid and efficient enzymatic digestion of cell walls, spheroplast lysis and complete protein precipitation, followed by DNA precipitation. XIT™ Genomic DNA from Yeast kit protocol is designed to use 1 ml overnight culture; however, the protocol can be easily adapted for larger tissue sample sizes.
G-Biosciences' XIT™ Mitochondrial DNA kit combines G-Biosciences FOCUS™ Mitochondria technology and XIT™ DNA isolation technology in a single kit. The kit uses subcell buffers to lyse cells and animal tissues and remove cellular proteins and nuceli from a highly enriched, intact and active mitochondrial fraction. The mitochondria are then lyzed with the included lysis buffer. The lysate is treated with G-Biosciences' LongLife™ Proteinase K to degrade the proteins, which are then precipitated and the mitochondrial DNA is isolated with alcohol precipitation. The kit protocol also contains protocols for isolation of mitochondra from soft (brain and liver) and hard (cardiac and skeletal muscle) tissue.
Ordering information: Accessories available.