SWIFT™ Western Blotting One Hour Detection System, G-Biosciences
Supplier: G-Biosciences
786-679 786-158
CA71003-234EA 501.56 CAD
CA71003-234 CA89167-736
SWIFT™ Western Blotting One Hour Detection System, G-Biosciences
Blotting Apparatus
G-Biosciences' SWIFT™ Western Diluent is a unique solution that reduces the blocking and antibody incubations on Western blot membranes from >4 hours to <60 minutes
An added advantage is that G-Biosciences' SWIFT™ Western Diluent is designed to be used with any combination of primary and secondary antibodies, unlike other commercial kits that limit researcher’s to rabbit or mouse primary antibodies. Compatible with wet and semi-dry transfer systems and with all automated transfer systems on the market.
G-Biosciences also offers the SWIFT™ Western Blotting System for addedMore Product Information