Sample Vials, Clear Borosilicate Glass, Screw-Thread
Supplier: Qorpak
GLA-07794 GLA-07796 GLC-07863 GLC-01022 GLA-08774 GLC-01014 GLC-01042 GLC-01027 GLC-01050 GLC-01029 GLC-00981 GLC-00985 GLC-05165 GLC-00994 GLC-04924 GLC-01009 GLC-01001 GLC-04995 GLC-04881 GLC-01007 GLC-04882 GLC-05182 GLC-04883 GLC-04947 GLC-04884 GLC-04835 GLC-05080 GLC-00996 GLC-00983 GLC-00987 GLC-04879 GLC-00998 GLC-04880 GLC-01011 GLC-04893 GLC-06160 GLC-00991 GLC-00995 GLA-00786 GLA-00787 GLA-00789 GLA-00790 GLC-01008 GLC-00978 GLC-00980 GLC-00988 GLC-00993 GLC-01002 GLC-00999 GLC-01006 GLA-00794 GLA-00791 GLA-00793 GLC-00982 GLC-00986 GLC-01003 GLC-01048 GLC-01016 GLA-00798 GLC-01015 GLC-01039 GLC-00984 GLC-06064 GLC-01021 GLC-05185 GLC-01031 GLC-08764 GLC-08765 GLA-07790 242633 242636 283154 GLC-07467 GLC-01013 GLC-09361 GLC-09036 GLC-07865 GLC-06994 GLC-00990 GLC-07367 GLC-13072 GLC-06849 GLC-05186 GLC-09161 GLC-09126 GLA-00788 GLA-00792 GLC-01020 GLC-07876
11311-214CS 351.54 CAD
11311-214 11311-216 11311-218 11311-514 11311-648 11311-650 16054-208 16054-212 16054-214 16054-222 16054-224 16087-042 16087-044 16087-048 16087-052 16087-056 16087-058 16087-060 16087-062 16087-064 16087-066 16087-068 16087-070 16087-072 16087-074 16087-076 16087-078 16087-088 16087-090 16087-122 16087-126 16087-128 16151-995 16218-124 16218-126 16218-128 16218-130 16218-132 16218-134 16218-136 18087-086 35100-129 36319-420 66009-546 66009-548 66009-550 66009-551 66009-552 66009-553 66009-554 66009-555 66009-556 66009-557 66009-559 66009-560 66009-561 66009-562 66009-563 66009-565 66009-566 66009-567 66009-575 66018-574 66018-665 66175-515 66175-520 89176-202 89176-854 93001-566 66009-558 93001-640 89210-118 89210-120 82020-162 10798-850 10798-852 10798-854 75832-344 76170-370 76170-374 76209-748 76403-540 76487-216 76487-218 76487-220 76487-222 76487-236 76487-238 76487-240 76487-242 76487-244 76487-352 76487-354 76487-356 76539-342 76539-344 76539-360 76539-362
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Vials
Clear, flint screw-thread sample vials offer maximum visibility.
  • Available with a variety of caps, or as vials only, to meet specific requirements
  • Pre-cleaned options save time and money
These general-purpose glass vials are made of Type I Borosilicate glass, providing exceptional resistance to heat shock and chemical leaching.