Protein Transfer Buffers, G-Biosciences
Supplier: G-Biosciences
786-019 786-423
CA82021-236EA 392.66 CAD
CA82021-236 CA95029-228
Protein Transfer Buffers, G-Biosciences
Buffers Electrophoresis Buffers Transfer Buffers
G-Biosciences' Efficient™ Western Transfer Buffer [20X] achieves increased protein transfer efficiency
G-Biosciences' High Molecular Weight Transfer Buffer [5X] is based on the Efficient™ Western Transfer Buffer and designed to facilitate the transfer of notoriously difficult high molecular weight proteins (>70kDa) during Western blotting. G-Biosciences' buffer is offered as a 1 liter supply of a 5X concentrated solution.
This ready-to-use transfer buffer is prepared for optimal conductivity and efficient protein transfer, without generating excessive heat or transfer distortion.
Ordering information: Accessories available.