Safety Coated Glass Jugs, Amber
Supplier: Qorpak
259523 259410 323205
10770-316CS 270.87 CAD
10770-316 10770-320 10770-402 76848-012
Safety Coated Glass Jugs, Amber
Bottles Carboys
Safety coated amber jugs with molded finger grips are the perfect storage solution for light sensitive products.
  • Amber glass protects against harmful UV rays
  • Jugs are slip and impact resistant, making them ideal for handling volatile substances
  • Choose jugs only, or jugs with attached Black Phenolic F217 and PTFE lined caps