Bottles, Cylinder, Natural High-Density Polyethylene, Narrow Mouth, Qorpak®
Supplier: Qorpak
PLA-03220 PLC-04831 PLA-03224 PLA-03214 PLC-03389 PLC-03427 PLA-03208 PLC-03401 PLA-03211 PLC-03435
16054-256CS 171.95 CAD
16054-256 16087-139 16087-289 16196-349 16218-155 36319-171 36319-180 36319-182 93001-758 89204-886 89204-888 89204-890 89204-894 89204-896 89204-906 89204-893 89204-911 76487-176
Bottles, Cylinder, Natural High-Density Polyethylene, Narrow Mouth, Qorpak®
Bottles Plastic Bottles
Natural HDPE commercial cylinder bottles are translucent and impact resistant.
They provide an excellent moisture barrier. Ideal for pharmaceutical applications. Narrow mouth cylinders are perfect for handling liquid samples in the lab. The shoulder style allows you to pour small quantities without spilling. HDPE bottles are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping.
Ordering information: Bottles 36319-180 and 36319-182 are supplied pre-cleaned for volatiles. Bottle 36319-171 is supplied pre-cleaned and certified for volatiles. Bottles 93001-754, 93001-758 and 76487-176 are supplied pre-cleaned and certified for metals. Bottle TNPLC09178 is supplied with Vacuum and Ionization treatment.