Safety-Coated Standard Wide Mouth Bottles, Clear, Qorpak®
Supplier: Qorpak
GLC-07366 GLC-02302 GLC-05729 GLC-05731 GLC-02247 GLC-02245 GLC-07364 262201
11310-996CS 397.55 CAD
11310-996 11311-113 11311-116 16054-253 16054-255 16152-340 16152-342 16193-916 16193-919 93000-924 89204-960 10847-232
Safety-Coated Standard Wide Mouth Bottles, Clear, Qorpak®
Bottles Glass Bottles
The wide opening on safety coated clear standard wide mouth bottles make them the ideal general purpose bottle for solid sampling.
The clear plastisol coating reduces the threat of breakage or lab mishaps.
Ordering information: Bottles 16054-253 is supplied pre-cleaned for volatiles. Bottle 93000-924 is supplied vacuum treated and ionized.