Safety-Coated Medium Round Bottles, Clear, Wide Mouth, Qorpak®
Supplier: Qorpak
GLC-07365 GLA-00945 GLA-00944 GLC-02239 GLC-02236
11310-994CS 558.23 CAD
11310-994 11311-112 16194-900 16194-903 16194-906 16194-909
Safety-Coated Medium Round Bottles, Clear, Wide Mouth, Qorpak®
Bottles Glass Bottles
Reduce breakage and lab mishaps with these safety coated clear medium round bottles.
If the glass breaks, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal.