Electronic Balance Calibration Weights, ANSI/ASTM Class 3 Weight, Troemner
Supplier: Troemner
80861367 80861390 80861396 80861442 80861443 80861444 80861439 80861440 80861441 80861436 80861437 80861438 80861424 80861425 80861426 80861421 80861422 80861423 80861418 80861419 80861420 80861415 80861416 80861417 80861412 80861413 80861414 80861409 80861410 80861411 80861384 80861385 80861386 80861364 80861365 80861366 80861358 80861359 80861360 80861352 80861353 80861354 80861346 80861347 80861348 80861326 80861327
11301-000EA 1058.3 CAD
11301-000 11301-002 11301-004 14216-556 14216-558 14216-560 14216-568 14216-570 14216-572 14216-580 14216-582 14216-584 14216-592 14216-594 14216-596 14216-604 14216-606 14216-608 14216-616 14216-618 14216-620 14216-628 14216-630 14216-632 14216-640 14216-642 14216-644 14216-652 14216-654 14216-656 14216-676 14216-678 14216-680 14216-694 14216-696 14216-698 14216-706 14216-708 14216-710 14216-718 14216-720 14216-722 14216-730 14216-732 14216-734 14216-742 14216-744
Electronic Balance Calibration Weights, ANSI/ASTM Class 3 Weight, Troemner
Weights Calibration Weights
Precision-crafted of stainless steel, with a highly polished finish
Weights 5kg and below supplied in a rugged, precision-fit polypropylene case with handling glove.
Standard weights are supplied with a Statement of Accuracy. Where applicable, weights are supplied with a NIST/NVLAP Weight Calibration Certificate or a Traceable Certificate.
Weights 30kg to 5kg are flat on top with a recessed grip handle for easy handling and stacking. Weights 5kg to 1kg are cylindrical in shape with a groove near the top to allow for easy handling and gripping. Weights below 1kg are straightMore Product Information