Safety-Coated Boston Round Bottles, Clear, Qorpak®
Supplier: Qorpak
GLC-08926 GLA-00937 GLA-00939 GLC-02214 GLC-02231 GLC-02206 GLC-02216 GLC-02224 GLC-02219 GLC-02227 GLA-00936 GLA-04900 GLA-00935 GLC-02213
11311-500CS 354.4 CAD
11311-500 15001-118 16150-804 16150-805 16150-808 16150-809 16150-812 16150-815 16150-818 16150-821 16150-824 16150-827 16150-831 16150-833 16150-834 16150-836 16150-837 16150-800 89204-884 30617-128 30617-136 76407-412
Safety-Coated Boston Round Bottles, Clear, Qorpak®
Bottles Glass Bottles
Safety coated clear Boston round bottles offers maximum visibility and sample integrity.
The safety coating on these bottles protects the container from being affected by pressure build up. If the glass breaks, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal. These bottles are slip and impact resistant, making them the perfect bottle for handling volatile substances.