OIML Class F1 Precision Weight Sets
Supplier: Troemner
80780454 80780439 80780448 80780450 80780413 80780412 80780421 80780423 80780427
12766-024EA 19188.66 CAD
12766-024 12766-046 12766-056 12766-060 12766-374 12766-376 12766-386 12766-390 12766-394
OIML Class F1 Precision Weight Sets
Weights Calibration Weights
Troemner OIML Calibration Weights conform to the standards of the Organisation Internationale de Metrologie Legale International Recommendation (OIML R 111)
Weights 1 g and above are two-piece construction. Troemner OIML Calibration Weights 1 g and larger are cylindrical in shape with a tapered neck for easy gripping. They are perfectly flat on top to stack easily on center. Weight bottoms are slightly recessed to expose the smallest possible area to wear.
They are manufactured from the finest stainless steels available including Troemner Alloy 8 Stainless Steel that has lowMore Product Information