OIML Class E2 Precision Weight Sets, Troemner
Supplier: Troemner
80780396 80780394 80780385 80780400
12766-134EA 11486.82 CAD
12766-134 12766-138 12766-148 12766-196
OIML Class E2 Precision Weight Sets, Troemner
Weights Calibration Weights
Troemner OIML Calibration Weights conform to the standards of the Organisation Internationale de Metrologie Legale International Recommendation (OIML R 111)
Weights 1 g and above are one-piece construction. Troemner OIML Calibration Weights 1 g and larger are cylindrical in shape with a tapered neck for easy gripping. They are perfectly flat on top to stack easily on center. Weight bottoms are slightly recessed to expose the smallest possible area to wear.
They are manufactured from the finest stainless steels available including Troemner Alloy 8 Stainless Steel that has low magnetic permeability, uniform consistency and high resistance to corrosion.