OIML Class F1 Precision Weights, Troemner
Supplier: Troemner
80780338 80780339 80780351 80780350 80780349 80780348 80780347 80780346 80780345 80780344 80780343 80780342 80780341 80780340 80780337 80780336 80780335 80780334 80780333 80780332 80780331 80780324 80780323 80780322 80780321 80780320 80780319 80780318 80780317 80780316 80780315 80780314 80780313 80780312 80780311 80780310 80780309 80780308 80780307 80780326 80780325 80780327
12765-996EA 297 CAD
12765-996 12765-998 12766-000 12766-002 12766-004 12766-006 12766-008 12766-010 12766-012 12766-014 12766-016 12766-018 12766-020 12766-022 12766-028 12766-030 12766-032 12766-034 12766-036 12766-038 12766-040 12766-336 12766-338 12766-340 12766-342 12766-344 12766-346 12766-348 12766-350 12766-352 12766-354 12766-356 12766-358 12766-360 12766-362 12766-364 12766-366 12766-368 12766-370 12766-400 12766-402 12766-404
OIML Class F1 Precision Weights, Troemner
Weights Calibration Weights
Troemner OIML Calibration Weights conform to the standards of the Organisation Internationale de Metrologie Legale International Recommendation (OIML R 111)
Weights 1 g and above are two-piece construction. Troemner OIML Calibration Weights 1 g and larger are cylindrical in shape with a tapered neck for easy gripping. They are perfectly flat on top to stack easily on center. Weight bottoms are slightly recessed to expose the smallest possible area to wear.
They are manufactured from the finest stainless steels available including Troemner Alloy 8 Stainless Steel that has lowMore Product Information