Ammonium persulfate (APS), powder for electrophoresis

Supplier: G-Biosciences
Danger

Synonyms: Ammonium persulfatePeroxidisulphuric acid diammonium saltAmmonium persulphatePeroxidisulfuric acid diammonium salt

RC-017 RC-017
CA95029-562EA 285.27 CAD
CA95029-562 95029-562
Ammonium persulfate (APS), powder for electrophoresis
Ammonium peroxodisulphate (APS)

G-Biosciences offers Ammonium Persulfate (APS), the catalyst for the polymerization of polyacrylamide gels, in the form of ready-to-use tablets (100 count) or as a powder (100 g)


For the tablets, simply add 1 tablet to 1.5 mL ultra pure water for a 10% solution.

Formula: (NH₄)₂S₂O₈
MW: 228.2 g/mol
Melting Pt: 120 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://ca.vwr.com/stibo/web/std.lang.all/06/39/16640639.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR