Gel Drying Solution for Protein Gels, G-Biosciences
Supplier: G-Biosciences
786-684 786-685 786-686
CA89167-896EA 240.18 CAD
CA89167-896 CA89167-898 CA89167-900
Gel Drying Solution for Protein Gels, G-Biosciences
Electrophoresis Gels Precast Gels
G-Biosciences ready-to-use gel drying solution effectively and evenly dries protein electrophoresis gels by the mechanism of passive evaporation and is suitable for permanent gel storage and densitometry.
The gel drying solution regulates the rate of drying and prevents the cracking of the gel. It is compatible with all drying membranes, including cellophane, and is supplied in three convenient sizes.