Economical Individual Weights, Troemner
Supplier: Troemner
30391015 30391012 30391009 30391006 30391003 30391000 30391014 30391011 30391008 30391005 30391002 30390999 30391013 30391010 30391007 30391004 30391001 30391016 30391017 30391018
97044-294EA 45.68 CAD
97044-294 97044-296 97044-298 97044-300 97044-302 97044-304 97044-306 97044-308 97044-310 97044-312 97044-314 97044-316 97044-318 97044-320 97044-322 97044-324 97044-326 10754-344 10754-346 10754-348
Economical Individual Weights, Troemner
Weights Calibration Weights
Individual weights are available from 2 kg to 1 mg.
Weights 1 g and above are carefully manufactured of stainless steel and are cylindrical in shape for easy gripping. Weights 1 g to 100 g are made from one-piece construction and are adjusted during manufacture by removing material from the bottom of the weight.
Weights 200 g through 2 kg have an adjusting cavity on the top that is sealed with a plug. Economical stainless steel weights 1 g and above are perfectly flat on top to stack easily on center.
Weights 500 mg and lower are made of sheet metal. They feature one-piece construction with one side turned up to make them easy to handle with forceps. The 500 mg and 200 mg weights are made of stainless steel while weights 100 mg and below are made of aluminum.
Certifications: Weights are adjusted to tolerances specified by ANSI/ASTM E617 Class 7.