Microliter™ and Gastight® Manual On Column GC Injection Syringes, Hamilton Company
Supplier: Hamilton
80331 80386 87402 87404 87405
60374-032EA 205.24 CAD
60374-032 60374-034 60373-480 89221-306 89221-308
Microliter™ and Gastight® Manual On Column GC Injection Syringes, Hamilton Company
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Each is manufactured to achieve the highest level of accuracy and precision possible. These syringes are designed to work with On Column gas chromatography (GC) injectors. Syringes in this category are either 700 or 7000 series Microliter™ or 1700 series Gastight® syringes. Microliter™ syringes have a stainless steel plunger that is hand fitted to the glass barrel and are designed for liquid sampling. Gastight® syringes have a PTFE plunger tip that creates tight connection between theMore Product Information