Troemner Alloy 8 Precision Analytical Weight Sets, Class 3, Troemner
Supplier: Troemner
80859289 80859292 80859291 80859334 80859336 80859335 80859319 80859321 80859320 80859386 80859388 80859387 80859263 80859265 80859264 80859478 80859480 80859479 80859250 80859251 80859252 80859223 80859224 80859225 80859197 80859198 80859199 80859167 80859168 80859169 80859141 80859142 80859143 80859115 80859116 80859117 80859089 80859090 80859091
12622-250EA 1803.35 CAD
12622-250 12622-251 12622-252 12626-270 12626-271 12626-272 12627-170 12627-171 12627-172 12627-175 12627-176 12627-177 12627-227 12627-228 12627-229 12765-898 12765-900 12765-902 14216-820 14216-822 14216-824 14216-844 14216-846 14216-848 14216-862 14216-864 14216-866 14216-874 14216-876 14216-878 14216-886 14216-888 14216-890 14216-910 14216-912 14216-914 14216-928 14216-930 14216-932
Troemner Alloy 8 Precision Analytical Weight Sets, Class 3, Troemner
Weights Calibration Weights
Highly polished kilogram and gram weights made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage
Fractional weights 500 mg to 5 mg are made of stainless steel (30 mg weights are made of aluminum); fractional weights 3mg and below are made of aluminum. Weights 500 g and smaller are supplied in rugged polypropylene cases. Sets that include weights 1 kg and larger are supplied in oak, aluminum, or rugged plastic cases. All cases are fitted with individual compartments and marked with the accuracy class and serial number. A clear fitted cover secures the fractional weights within the case. Each case includes contoured weight forceps. Analytical weights 1g and above are manufactured with adjusting cavities.
This exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to –195.5°C (–320°F).
Certifications: Standard weight sets are supplied with a Statement of Accuracy. Where applicable, weight sets are supplied with a NIST/NVLAP Weight Calibration Certificate or a Traceable Certificate.