Troemner Alloy 8 Precision Analytical Weight Sets, Class 2, Troemner
Supplier: Troemner
80859260 80859286 80859315 80859330 80859383 80859261 80859287 80859317 80859332 80859384 80859262 80859288 80859318 80859333 80859385 80859475 80859477 80859476 80859247 80859248 80859249 80859220 80859221 80859222 80859194 80859195 80859196 80859164 80859165 80859166 80859138 80859139 80859140 80859112 80859113 80859114 80859086 80859087 80859088
12666-000EA 1975.74 CAD
12666-000 12666-002 12666-004 12666-006 12666-009 12666-020 12666-022 12666-024 12666-026 12666-028 12666-030 12666-032 12666-034 12666-036 12666-038 12765-892 12765-894 12765-896 14216-814 14216-816 14216-818 14216-838 14216-840 14216-842 14216-856 14216-858 14216-860 14216-868 14216-870 14216-872 14216-880 14216-882 14216-884 14216-904 14216-906 14216-908 14216-922 14216-924 14216-926
Troemner Alloy 8 Precision Analytical Weight Sets, Class 2, Troemner
Weights Calibration Weights
Highly polished kilogram and gram weights made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage
Fractional weights 500 mg to 5 mg are made of stainless steel (30 mg weights are made of aluminum); fractional weights 3mg and below are made of aluminum. Weights 500 g and smaller are supplied in rugged polypropylene cases. Sets that include weights 1 kg and larger are supplied in oak, aluminum, or rugged plastic cases. All casesMore Product Information