FreeZone® Freeze Dry Glassware, Labconco
Supplier: Labconco
7551000 7554200 7554400 7554600 7554900 7552000 7552200 7552400 7556000 7556200 7556400 7557000 7557200 7557400 7557600 7568600 7570610 7570810 7571010 7571210 7573010 7573210 7573410 7573610 7573810 7574010 7574210 7575010 7575210 7557800 7558000 7550000 7550200 7550400 7550600 7550800 7550900 7554000 7555000 7568000 7568200 7568400 7572710 7554800 7557900
26674-250EA 1098.1 CAD
26674-250 26674-415 26674-420 26674-425 26674-440 26674-522 26674-524 26674-526 26674-528 26674-530 26674-532 26674-534 26674-536 26674-538 26674-540 26675-104 26675-570 26675-572 26675-574 26675-576 26675-685 26675-687 26675-689 26675-691 26675-693 26675-695 26675-697 26675-699 26675-701 26675-872 30153-694 26674-140 26674-145 26674-156 26674-178 26674-214 26674-236 26674-350 26674-451 26675-068 26675-079 26675-090 26675-680 10148-034 10148-042
FreeZone® Freeze Dry Glassware, Labconco
Freeze Dryers
FreeZone® Freeze Dry Systems were designed to provide simplicity of use and convenience when freeze drying samples for the laboratory.
  • A variety of accessories, including glassware, are available for these units
  • Glassware offered includes flasks, ampules, serum bottles and vials
Labconco Ampules are fabricated of highest quality borosilicate glass for strength and durability
Serum Bottles are perfect for long term storage of freeze dried samples. Labconco serum bottles are specifically designed for lyophilization applications. Their uniform thin wall construction ensures even freezing and drying. Serum Bottles are ideal containers for use in the FreeZone Stoppering Tray Dryer and also connect to valve ports on drying chambers and manifolds.
Adapters connect Lyph-Lock® Flasks to the valve ports on a drying chamber or manifold of a freeze dryer. These adapters have a 90° bend.
Lyph-Lock® Flasks come in a variety of sizes to suit different sized samples. Flasks should be filled to no more than one-third of their volume so that maximum surface area is achieved and efficient lyophilization is ensured. A complete Lyph-Lock Flask Flask includes a glass top and bottom and a rubber ring seal. Tops, bottoms and seals are available separately as replacement components. Adapters are required to attach flasks to freeze dry valve ports.
Ordering information: For Fast-Freeze Flasks in clear and amber-colored glass, contact your Avantor representative for more information.