Nunc® Multidishes, Non-treated, Thermo Scientific
Supplier: Thermo Fisher Scientific
150239 150200 144530 150787 179820
CA73521-138CS 629.88 CAD
CA73521-138 CA73521-140 CA73521-142 CA73521-144 CA73521-356
Nunc® Multidishes, Non-treated, Thermo Scientific
Culture Dishes Cell Culture Dishes
These multi dishes are non-treated and can be used for the suspension of cell cultures
The dishes have raised well rims which lowers the risk of cross-contamination. They also have flat bottom wells that allow for easy viewing of cultures. Lids with notched corners for a good fit are included. The lids help with single position orientation and cross contamination prevention. The multi dishes are made of sterile polystyrene.
Order Now
SPECIFICATIONS
Environmentally Preferable