Gastight® 1700 Series Gel Loading Syringes, Hamilton Company
Supplier: Hamilton
84505 84500 84501 84504 84502 84503
60374-038EA 172.1 CAD
60374-038 60374-048 89221-282 89221-288 89221-284 89221-286
Gastight® 1700 Series Gel Loading Syringes, Hamilton Company
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Each is manufactured to achieve the highest level of accuracy and precision possible. These syringes are available in single and multi-channel formats. Multi-Channel Gel Loading syringes are designed for multiplex DNA sequencing methodologies. You can rapidly transfer eight samples to sequencing gels, microwell plates or nylon membranes. It accurately dispenses from 0.2 μl to 10 μl of sample per syringe simultaneously. The 9 mm needle spacing and the three needle diameters optimize sample transfers. There is an adjustable volume stop that allows faster, more accurate reloading and excellent reproducibility. Adjustable Multi-Channel Gel Loading syringes simplify and speed sample loading of 96-lane gels. They are compatible with 96-lane ABI 377 PRISM® DNA sequencing instrument. They have an easy-to-use design for needle spacing adjustment. The removable needle designs are available with 0.2 mm. The adjustable Gel Loading syringe (AGLS) allows quick, easy transfer and loading of DNA samples. Eight samples at a time can be transferred directly from a microwell plate with 9 mm spacing to a 96-lane gel with 10.8 mm spacing. Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal.
Cemented Needle (N)- For low volume syringes the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable. Special Cemented Needle (SN) - For low volume syringes the special needle terminations are the same as the Cemented Needle terminations except they allow for a variety of user-defined gauges, lengths, and point styles to be attached. Removable Needle (RN)- The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life. Needle Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 0.2, 0.3, 0.4 mm.