Microliter™ Carbon Analyzer Syringes, Hamilton Company
Supplier: Hamilton
80550 80770 80660 80551
60377-302EA 235.2 CAD
60377-302 72310-318 72310-320 89221-266
Microliter™ Carbon Analyzer Syringes, Hamilton Company
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Each is manufactured to achieve the highest level of accuracy and precision possible. These syringes are designed for water analysis with Total Organic Carbon. Syringes in this category are 700 series Microliter™ syringes. They have a stainless steel plunger hand fitted to the glass barrel and are designed for liquid sampling.
These syringes have a carbon analyzer (CA) termination.
Carbon Analyzer (CA) - This is a special type of cemented needle termination. The needle is a metal luer that is cemented onto the syringe barrel. This termination is exclusively used on Carbon Analyzer syringes only.
Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 24, 26