Nunc® MicroWell™ 96-Well Optical-Bottom Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
265300 164588 164590 160376
CA73520-168CS 4307.02 CAD
CA73520-168 CA73520-174 CA73520-178 CA73520-750
Nunc® MicroWell™ 96-Well Optical-Bottom Plates, Thermo Scientific
Microplates Immunology Microplates and Strips
The Plates are optimized for fluorescence and luminescence in cell culture and microscopic applications
The coverglass bottom has minimum light scatter and low autofluorescence, ensuring accurate results due to higher signal-to-noise ratio.