InLab® Intelligent Sensor Management (ISM®) pH/Conductivity Electrodes, METTLER TOLEDO
Supplier: Mettler Toledo
51344155 51344211 51344102 51344104 51344055 51344072 51344172 51344163 51344110 51344114 51344116 51343156 30014096 30014092 30014093 30014094
CA97025-876EA 1530.25 CAD
CA97025-876 CA97025-878 CA97025-870 CA97025-872 CA97025-874 CA97025-880 CA97025-882 CA97025-884 CA97025-888 CA97025-890 CA97025-892 CA97044-112 CA89207-248 CA89207-252 CA89207-254 CA89207-256
InLab® Intelligent Sensor Management (ISM®) pH/Conductivity Electrodes, METTLER TOLEDO
Electrodes Multi-Parameter Electrodes
ISM® is a sophisticated safety concept that leaves nothing to chance.
  • Intelligent sensor management system (ISM®) internal reference
  • Calibration automatically transferred between compatible METTLER TOLEDO meters
  • Certificate can be sent to printer or PC
  • If the electrode comes with a cable, it will be listed in the order section below
The ISM® technology is compatible with following METTLER TOLEDO Meters: SevenExcellence™, SevenDirect™, SevenGo pro™, SevenGo Duo pro™, and SevenCompact™ (discontinued in June 2022).
The electrode certificate with theMore Product Information