Mammalian GenLysate™ Rabbit Total Protein Lysates, G-Biosciences
Supplier: G-Biosciences
NLB-01 NLB-03 NLB-04 NLB-08 NLB-10 NLB-21 SLB-1001 SLB-1002 SLB-1003
CA82021-692EA 582.14 CAD
CA82021-692 CA82021-696 CA82021-700 CA82021-704 CA82021-708 CA82021-712 CA82023-116 CA82023-120 CA82023-124
Mammalian GenLysate™ Rabbit Total Protein Lysates, G-Biosciences
Cells
G-Biosciences' GenLysates™ are total protein tissue and cell lysates that are extracted from the tissue or cell of interest. To inhibit proteolysis, the GenLysates™ are prepared in a lysis buffer containing a complete protease inhibitor cocktail. Each lysate lot is tested by electrophoresis and Western blot analysis.
G-Biosciences' GenLysate™ total protein lysates are ready to use for SDS-PAGE and Western blotting purposes
G-Biosciences' GenLysate™ total protein lysates are offered as a 150 µg lyophilized protein per vial and, following reconstitution in DI water, are ready for use. G-Biosciences offers GenLysate™ in a wide variety of different species and tissues. Species include: human (both normal tissue and tumor tissue lysates available), mouse, rat, macaque (primate), rabbit, pig, plant-related, bacteria, insect, bird, frog, fish, marsupial, earthworm, and yeast.
The total proteins are being extracted from whole tissue or cells following the method of Laemmli. The proteins are extracted in a denaturing buffer containing 62 mM Tris·HCl and 2% SDS with G-Biosciences' ProteaseARREST™ protease inhibitor cocktail. This minimizes proteolytic damage to the proteins.