Mammalian GenLysate™ Rat Total Protein Lysates, G-Biosciences
Supplier: G-Biosciences
NLR-01 NLR-02 NLR-03 NLR-04 NLR-05 NLR-06 NLR-07 NLR-08 NLR-09 NLR-10 SLR-0210 SLR-0211 SLR-0212 SLR-022 SLR-023 SLR-024 SLR-025 SLR-026 SLR-027 SLR-028 SLR-029 SLR-041 SLR-042 SLR-1001 SLR-1002 SLR-1003 SLR-021
CA82021-874EA 529.57 CAD
CA82021-874 CA82021-878 CA82021-882 CA82021-888 CA82021-894 CA82021-898 CA82021-902 CA82021-906 CA82021-910 CA82021-914 CA82022-224 CA82022-228 CA82022-232 CA82022-238 CA82022-242 CA82022-246 CA82022-250 CA82022-254 CA82022-258 CA82022-262 CA82022-266 CA82022-270 CA82022-274 CA82022-278 CA82022-282 CA82022-286 CA95029-336
Mammalian GenLysate™ Rat Total Protein Lysates, G-Biosciences
Cells
G-Biosciences' GenLysates™ are total protein tissue and cell lysates that are extracted from the tissue or cell of interest. To inhibit proteolysis, the GenLysates™ are prepared in a lysis buffer containing a complete protease inhibitor cocktail. Each lysate lot is tested by electrophoresis and Western blot analysis.
G-Biosciences' GenLysate™ total protein lysates are ready to use for SDS-PAGE and Western blotting purposes
G-Biosciences' GenLysate™ total protein lysates are offered as a 150 µg lyophilized protein per vial and, following reconstitution in DI water, are ready for use. G-Biosciences offers GenLysate™ in a wide variety of different species and tissues. Species include: human (both normal tissue and tumor tissue lysates available), mouse, rat, macaque (primate), rabbit, pig, plant-related, bacteria, insect, bird, frog, fish, marsupial, earthworm, and yeast.
The total proteins are being extracted from whole tissue or cells following the method of Laemmli. The proteins are extracted in a denaturing buffer containing 62 mM Tris·HCl and 2% SDS with G-Biosciences' ProteaseARREST™ protease inhibitor cocktail. This minimizes proteolytic damage to the proteins.