Nunc® 96-Well Polystyrene Conical Bottom Microwell Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
249952 249940 249935
CA73520-134CS 1815.18 CAD
CA73520-134 CA89027-038 CA89027-036
Nunc® 96-Well Polystyrene Conical Bottom Microwell Plates, Thermo Scientific
Microplates Cell Culture Plates
Nunc® 96-well polystyrene conical plates are optimized for robotics, automated readers and liquid handling systems
The plates are clear which makes them ideal for optical measurements. The lids in the package come with rings which prevent cross contamination. The well shape of these plates maximizes sample recovery.
Certifications: All microplates manufactured by Nunc® meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)