4-tert-Butylbenzylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L20394-06
L20394-14
CAAAAL20394-06EA
105.86
CAD
CAAAAL20394-06
CAAAAL20394-14
4-tert-Butylbenzylamine 98%
4-tert-Butylbenzylamine
Purity: 98%
CAS No.: 39895-55-1
Molecular Formula: C11H17N
Formula Weight: 163.26
Flash Point: 107°(224°F)
Refractive Index: 1.5210
Boiling Point: 124°/16mm
Beilstein Reference: 2961797
UN No.: 2735
MDL No.: MFCD00040754
Formula:
C₁₁H₁₇N Boiling Pt: 124 °C/16 mm Density: 0.927 Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040754 CAS Number: 39895-55-1 EINECS: 254-681-6 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...